Assay Design
Gene Transcripts
- Gene Symbol
- nas-19
- Entrez Gene ID
-
186926
- Gene Name
- Zinc metalloproteinase nas-19
- Gene Aliases
-
CELE_K03B8.5
- Chromosome Location
-
Chr.V: 11401091 - 11402710 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073492.4 |
NP_505893.2 |
1 - 2 |
320 |
89 |
View Full Product Details