Assay Design



Gene Transcripts

Gene Symbol
nhr-68
Entrez Gene ID
179589
Gene Name
protein-coding
Gene Aliases
CELE_H12C20.3
Chromosome Location
Chr.V: 11573440 - 11575839 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380769.2 NP_001366648.1 2 - 3 423 100

View Full Product Details