Assay Design
Gene Transcripts
- Gene Symbol
- glb-15
- Entrez Gene ID
-
185260
- Gene Name
- GLoBin related
- Gene Aliases
-
CELE_F35B12.8
- Chromosome Location
-
Chr.V: 11616805 - 11618273 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073546.6 |
NP_505947.2 |
5 - 6 |
562 |
123 |
View Full Product Details