Assay Design
Gene Transcripts
- Gene Symbol
- lgc-44
- Entrez Gene ID
-
179600
- Gene Name
- Ligand-Gated ion Channel
- Gene Aliases
-
CELE_F46F3.2
- Chromosome Location
-
Chr.V: 11669338 - 11672597 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073553.6 |
NP_505954.2 |
7 - 8 |
1506 |
77 |
View Full Product Details