Assay Design



Gene Transcripts

Gene Symbol
tre-3
Entrez Gene ID
179604
Gene Name
protein-coding
Gene Aliases
CELE_W05E10.4
Chromosome Location
Chr.V: 11709769 - 11715903 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269432.3 NP_001256361.1 9 - 10 1647 66
NM_001269433.3 NP_001256362.1 1 - 2 51 66

View Full Product Details