Assay Design



Gene Transcripts

Gene Symbol
clh-6
Entrez Gene ID
179655
Gene Name
Chloride channel protein
Gene Aliases
CELE_R07B7.1
Chromosome Location
Chr.V: 12060233 - 12063368 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073621.5 NP_506022.1 6 - 7 1397 112
GenBank mRNA AF319616.1 - 6 - 7 1419 112
AF328908.1 - 6 - 7 1428 112

View Full Product Details