Assay Design
Gene Transcripts
- Gene Symbol
- che-11
- Entrez Gene ID
-
179666
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_C27A7.4
- Chromosome Location
-
Chr.V: 12144244 - 12150707 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073646.7 |
NP_506047.5 |
12 - 13 |
2232 |
108 |
View Full Product Details