Assay Design
Gene Transcripts
- Gene Symbol
- srj-4
- Entrez Gene ID
-
182952
- Gene Name
- Serpentine Receptor, class J
- Gene Aliases
-
CELE_C27A7.7
- Chromosome Location
-
Chr.V: 12163206 - 12164535 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073650.1 |
NP_506051.1 |
1 - 2 |
253 |
90 |
View Full Product Details