Assay Design
Gene Transcripts
- Gene Symbol
- ugt-56
- Entrez Gene ID
-
188073
- Gene Name
- Putative UDP-glucuronosyltransferase ugt-56
- Gene Aliases
-
CELE_T04H1.8
- Chromosome Location
-
Chr.V: 12258982 - 12261215 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073673.6 |
NP_506074.2 |
5 - 6 |
1091 |
99 |
View Full Product Details