Assay Design



Gene Transcripts

Gene Symbol
pals-39
Entrez Gene ID
183784
Gene Name
Protein containing ALS2cr12 (ALS2CR12) signature
Gene Aliases
CELE_C54D10.7
Chromosome Location
Chr.V: 12441663 - 12443854 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269488.3 NP_001256417.1 7 - 8 1095 69

View Full Product Details