Assay Design



Gene Transcripts

Gene Symbol
atn-1
Entrez Gene ID
179709
Gene Name
protein-coding
Gene Aliases
CELE_W04D2.1
Chromosome Location
Chr.V: 12478411 - 12484449 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269495.3 NP_001256424.1 3 - 4 564 112
NM_073727.5 NP_506128.1 4 - 5 782 112

View Full Product Details