Assay Design



Gene Transcripts

Gene Symbol
ril-2
Entrez Gene ID
179725
Gene Name
protein-coding
Gene Aliases
CELE_C14C10.3
Chromosome Location
Chr.V: 12596169 - 12599157 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028487.7 NP_001023658.1 2 - 3 180 66
NM_001028488.6 NP_001023659.1 2 - 3 182 66
NM_001269507.4 NP_001256436.1 2 - 3 186 66

View Full Product Details