Assay Design
Gene Transcripts
- Gene Symbol
- gpa-8
- Entrez Gene ID
-
179731
- Gene Name
- Guanine nucleotide-binding protein alpha-8 subunit
- Gene Aliases
-
CELE_F56H9.3
- Chromosome Location
-
Chr.V: 12638666 - 12647432 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073759.3 |
NP_506160.1 |
7 - 8 |
614 |
150 |
GenBank mRNA |
AY008131.1 |
- |
3 - 4 |
183 |
150 |
View Full Product Details