Assay Design
Gene Transcripts
- Gene Symbol
- his-74
- Entrez Gene ID
-
179734
- Gene Name
- Histone H3.3-like type 2
- Gene Aliases
-
CELE_W05B10.1
- Chromosome Location
-
Chr.V: 12659704 - 12660330 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073763.5 |
NP_506164.1 |
1 - 2 |
286 |
67 |
View Full Product Details