Assay Design
Gene Transcripts
- Gene Symbol
- ugt-3
- Entrez Gene ID
-
191174
- Gene Name
- UDP-GlucuronosylTransferase
- Gene Aliases
-
CELE_ZC455.3
- Chromosome Location
-
Chr.V: 12798308 - 12800271 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073802.3 |
NP_506203.2 |
4 - 5 |
1253 |
81 |
View Full Product Details