Assay Design



Gene Transcripts

Gene Symbol
srr-1
Entrez Gene ID
189285
Gene Name
protein-coding
Gene Aliases
CELE_W07G4.6
Chromosome Location
Chr.V: 13049730 - 13053735 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313070.4 NP_001299999.1 2 - 2 286 121
NM_001377607.2 NP_001364593.1 2 - 3 282 121

View Full Product Details