Assay Design
Gene Transcripts
- Gene Symbol
- eat-6
- Entrez Gene ID
-
179796
- Gene Name
- Sodium/potassium-transporting ATPase subunit alpha
- Gene Aliases
-
CELE_B0365.3
- Chromosome Location
-
Chr.V: 13126413 - 13129995 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073868.5 |
NP_506269.1 |
1 - 2 |
129 |
147 |
View Full Product Details