Assay Design
Gene Transcripts
- Gene Symbol
- odr-3
- Entrez Gene ID
-
179806
- Gene Name
- Guanine nucleotide-binding protein alpha-17 subunit
- Gene Aliases
-
CELE_C34D1.3
- Chromosome Location
-
Chr.V: 13246308 - 13249536 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073889.2 |
NP_506290.1 |
3 - 4 |
593 |
82 |
GenBank mRNA |
AY008142.1 |
- |
3 - 4 |
591 |
82 |
View Full Product Details