Assay Design



Gene Transcripts

Gene Symbol
uig-1
Entrez Gene ID
179808
Gene Name
protein-coding
Gene Aliases
CELE_F32F2.1
Chromosome Location
Chr.V: 13273157 - 13295978 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269559.3 NP_001256488.1 15 - 16 2709 90
NM_001269560.3 NP_001256489.1 15 - 16 2751 90
NM_001269563.3 NP_001256492.1 11 - 12 2160 90
NM_001269564.4 NP_001256493.1 7 - 8 1083 90
NM_001269565.3 NP_001256494.1 5 - 6 513 90
NM_001380797.1 NP_001367249.1 15 - 16 2691 90
NM_001380798.1 NP_001366647.1 15 - 16 2843 90

View Full Product Details