Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001269559.3 | NP_001256488.1 | 4 - 5 | 504 | 61 |
NM_001269560.3 | NP_001256489.1 | 4 - 5 | 546 | 61 | |
NM_001380797.1 | NP_001367249.1 | 4 - 5 | 486 | 61 | |
NM_001380798.1 | NP_001366647.1 | 4 - 5 | 638 | 61 |