Assay Design
Gene Transcripts
- Gene Symbol
- str-37
- Entrez Gene ID
-
183639
- Gene Name
- Seven TM Receptor
- Gene Aliases
-
CELE_C50B6.12
- Chromosome Location
-
Chr.V: 13327763 - 13329058 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073903.2 |
NP_506304.1 |
1 - 2 |
552 |
117 |
View Full Product Details