Assay Design
Gene Transcripts
- Gene Symbol
- srg-46
- Entrez Gene ID
-
185217
- Gene Name
- Serpentine receptor class gamma-46
- Gene Aliases
-
CELE_F32H5.5
- Chromosome Location
-
Chr.V: 13363715 - 13364728 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073913.2 |
NP_506314.2 |
1 - 2 |
136 |
128 |
View Full Product Details