Assay Design



Gene Transcripts

Gene Symbol
rdy-2
Entrez Gene ID
186178
Gene Name
protein-coding
Gene Aliases
CELE_F53F4.6
Chromosome Location
Chr.V: 13598331 - 13600982 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129478.2 NP_001122950.1 3 - 4 426 124
NM_001129479.2 NP_001122951.1 3 - 4 426 124

View Full Product Details