Assay Design
Gene Transcripts
- Gene Symbol
- letm-1
- Entrez Gene ID
-
179854
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F58G11.1
- Chromosome Location
-
Chr.V: 13661255 - 13664233 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073981.7 |
NP_506382.1 |
5 - 6 |
2266 |
63 |
View Full Product Details