Assay Design
Gene Transcripts
- Gene Symbol
- hlh-34
- Entrez Gene ID
-
179860
- Gene Name
- Helix-loop-helix 34
- Gene Aliases
-
CELE_T01D3.2
- Chromosome Location
-
Chr.V: 13705967 - 13707395 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073990.5 |
NP_506391.2 |
2 - 3 |
316 |
103 |
GenBank mRNA |
AF370362.1 |
- |
2 - 3 |
281 |
103 |
View Full Product Details