Assay Design
Gene Transcripts
- Gene Symbol
- ttr-21
- Entrez Gene ID
-
184253
- Gene Name
- TransThyretin-Related family domain
- Gene Aliases
-
CELE_F09F3.6
- Chromosome Location
-
Chr.V: 13853834 - 13855260 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074032.8 |
NP_506433.1 |
1 - 2 |
1041 |
99 |
View Full Product Details