Assay Design
Gene Transcripts
- Gene Symbol
- oac-3
- Entrez Gene ID
-
182290
- Gene Name
- NRF domain-containing protein
- Gene Aliases
-
CELE_C06B3.2
- Chromosome Location
-
Chr.V: 13899513 - 13902653 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074045.2 |
NP_506446.2 |
7 - 8 |
1014 |
124 |
View Full Product Details