Assay Design



Gene Transcripts

Gene Symbol
srxa-12
Entrez Gene ID
3564785
Gene Name
Serpentine Receptor, class XA
Gene Aliases
CELE_C06B3.13
Chromosome Location
Chr.V: 13926698 - 13929320 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026198.2 NP_001021369.1 6 - 7 1126 98

View Full Product Details