Assay Design
Gene Transcripts
- Gene Symbol
- srx-42
- Entrez Gene ID
-
182296
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_C06B3.11
- Chromosome Location
-
Chr.V: 13921030 - 13922404 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074054.2 |
NP_506455.2 |
4 - 5 |
447 |
106 |
View Full Product Details