Assay Design



Gene Transcripts

Gene Symbol
srxa-11
Entrez Gene ID
3565536
Gene Name
Serpentine Receptor, class XA
Gene Aliases
CELE_C06B3.12
Chromosome Location
Chr.V: 13924151 - 13925807 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026197.1 NP_001021368.1 4 - 5 504 83

View Full Product Details