Assay Design
Gene Transcripts
- Gene Symbol
- end-1
- Entrez Gene ID
-
179893
- Gene Name
- GATA-type domain-containing protein
- Gene Aliases
-
CELE_F58E10.2
- Chromosome Location
-
Chr.V: 13985128 - 13987074 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074074.3 |
NP_506475.1 |
2 - 3 |
396 |
75 |
View Full Product Details