Assay Design
Gene Transcripts
- Gene Symbol
- aip-1
- Entrez Gene ID
-
179898
- Gene Name
- Arsenite Inducible Protein
- Gene Aliases
-
CELE_F58E10.4
- Chromosome Location
-
Chr.V: 14012333 - 14013644 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074078.4 |
NP_506479.1 |
2 - 3 |
404 |
92 |
View Full Product Details