Assay Design



Gene Transcripts

Gene Symbol
gar-3
Entrez Gene ID
179962
Gene Name
protein-coding
Gene Aliases
CELE_Y40H4A.1
Chromosome Location
Chr.V: 14564819 - 14579719 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029064.4 NP_001024235.1 3 - 4 215 59
NM_001029065.5 NP_001024236.1 1 - 2 70 59
GenBank mRNA AF139093.2 - 3 - 4 237 59
AY221631.1 - 1 - 2 70 59

View Full Product Details