Assay Design



Gene Transcripts

Gene Symbol
pmp-3
Entrez Gene ID
179968
Gene Name
protein-coding
Gene Aliases
CELE_C54G10.3
Chromosome Location
Chr.V: 14646739 - 14653493 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269678.3 NP_001256607.1 3 - 4 500 142
NM_001269679.3 NP_001256608.1 2 - 3 402 142

View Full Product Details