Assay Design



Gene Transcripts

Gene Symbol
cng-1
Entrez Gene ID
180004
Gene Name
Cyclic nucleotide-binding domain-containing protein
Gene Aliases
CELE_F14H8.6
Chromosome Location
Chr.V: 15025731 - 15031473 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129446.4 NP_001122918.1 11 - 12 2147 142
NR_070019.1 12 - 13 1942 142
NR_070020.1 11 - 12 1819 142

View Full Product Details