Assay Design
Gene Transcripts
- Gene Symbol
- srw-16
- Entrez Gene ID
-
188394
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_T10H4.5
- Chromosome Location
-
Chr.V: 15273383 - 15274857 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074381.2 |
NP_506782.2 |
2 - 3 |
950 |
140 |
View Full Product Details