Assay Design
Gene Transcripts
- Gene Symbol
- srt-22
- Entrez Gene ID
-
191498
- Gene Name
- Serpentine Receptor, class T
- Gene Aliases
-
CELE_ZK1037.3
- Chromosome Location
-
Chr.V: 15314668 - 15315927 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074395.1 |
NP_506796.1 |
4 - 5 |
873 |
134 |
View Full Product Details