Assay Design
Gene Transcripts
- Gene Symbol
- str-23
- Entrez Gene ID
-
183826
- Gene Name
- Seven TM Receptor
- Gene Aliases
-
CELE_C55A1.5
- Chromosome Location
-
Chr.V: 15644875 - 15646478 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074490.1 |
NP_506891.1 |
3 - 4 |
537 |
135 |
View Full Product Details