Assay Design
Gene Transcripts
- Gene Symbol
- twk-34
- Entrez Gene ID
-
180054
- Gene Name
- TWiK family of potassium channels
- Gene Aliases
-
CELE_K06B4.12
- Chromosome Location
-
Chr.V: 15704960 - 15709592 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074505.5 |
NP_506906.3 |
8 - 9 |
1436 |
97 |
View Full Product Details