Assay Design



Gene Transcripts

Gene Symbol
str-27
Entrez Gene ID
188782
Gene Name
Seven TM Receptor
Gene Aliases
CELE_T23D5.1
Chromosome Location
Chr.V: 15734023 - 15735820 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322595.1 NP_001309461.1 3 - 4 494 86

View Full Product Details