Assay Design



Gene Transcripts

Gene Symbol
str-43
Entrez Gene ID
188788
Gene Name
Seven TM Receptor
Gene Aliases
CELE_T23D5.9
Chromosome Location
Chr.V: 15750932 - 15752149 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322596.1 NP_001309462.1 1 - 2 495 90

View Full Product Details