Assay Design
Gene Transcripts
- Gene Symbol
- cyp-35D1
- Entrez Gene ID
-
184495
- Gene Name
- CYtochrome P450 family
- Gene Aliases
-
CELE_F14H3.10
- Chromosome Location
-
Chr.V: 16069238 - 16071318 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074643.2 |
NP_507044.1 |
5 - 6 |
1380 |
131 |
View Full Product Details