Assay Design
Gene Transcripts
- Gene Symbol
- srxa-16
- Entrez Gene ID
-
185755
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_F44G3.5
- Chromosome Location
-
Chr.V: 16112992 - 16114337 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074655.2 |
NP_507056.2 |
2 - 3 |
373 |
102 |
View Full Product Details