Assay Design
Gene Transcripts
- Gene Symbol
- oac-15
- Entrez Gene ID
-
184335
- Gene Name
- Acyl_transf_3 domain-containing protein
- Gene Aliases
-
CELE_F11A5.8
- Chromosome Location
-
Chr.V: 16206849 - 16209937 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074687.3 |
NP_507088.1 |
2 - 3 |
405 |
114 |
View Full Product Details