Assay Design
Gene Transcripts
- Gene Symbol
- oac-7
- Entrez Gene ID
-
180096
- Gene Name
- NRF domain-containing protein
- Gene Aliases
-
CELE_C31A11.7
- Chromosome Location
-
Chr.V: 16309805 - 16314775 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074719.5 |
NP_507120.3 |
2 - 3 |
392 |
119 |
View Full Product Details