Assay Design
Gene Transcripts
- Gene Symbol
- srw-29
- Entrez Gene ID
-
184965
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_F26D2.11
- Chromosome Location
-
Chr.V: 16423099 - 16424528 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074750.2 |
NP_507151.2 |
3 - 4 |
915 |
124 |
View Full Product Details