Assay Design
Gene Transcripts
- Gene Symbol
- srr-3
- Entrez Gene ID
-
185351
- Gene Name
- Serpentine Receptor, class R
- Gene Aliases
-
CELE_F36D3.3
- Chromosome Location
-
Chr.V: 16506894 - 16508767 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074779.5 |
NP_507180.1 |
2 - 3 |
411 |
76 |
View Full Product Details