Assay Design
Gene Transcripts
- Gene Symbol
- srw-54
- Entrez Gene ID
-
189573
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_Y32B12C.2
- Chromosome Location
-
Chr.V: 16611335 - 16612703 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074801.1 |
NP_507202.1 |
2 - 3 |
465 |
94 |
View Full Product Details