Assay Design



Gene Transcripts

Gene Symbol
srw-54
Entrez Gene ID
189573
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_Y32B12C.2
Chromosome Location
Chr.V: 16611335 - 16612703 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074801.1 NP_507202.1 3 - 4 871 106
NR_023987.1 4 - 5 986 106

View Full Product Details