Assay Design
Gene Transcripts
- Gene Symbol
- srw-44
- Entrez Gene ID
-
184480
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_F14F8.6
- Chromosome Location
-
Chr.V: 16680281 - 16682656 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074822.1 |
NP_507223.1 |
4 - 5 |
954 |
195 |
View Full Product Details